Benzoyl-DL-methionine ≥99.0% (by titrimetric analysis)

Synonyms: Bz-DL-Met-OH

B0300-1G B0300-10G
TCB0300-001GEA 28.56 USD
TCB0300-001G TCB0300-010G
Benzoyl-DL-methionine ≥99.0% (by titrimetric analysis)
Benzoyl-DL-methionine

CAS Number: 4703-38-2
MDL Number: MFCD00026044
Molecular Formula: C12H15NO3S
Molecular Weight: 253.32
Purity/Analysis Method: 99.0% (T)
Form: Crystal
Melting point (°C): 152

Formula: C₁₂H₁₅NO₃S
MW: 253.32 g/mol
Melting Pt: 152 °C
MDL Number: MFCD00026044
CAS Number: 4703-38-2
https://us.vwr.com/stibo/web/std.lang.all/23/43/28932343.jpg

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR