3,5-Diphenyl-1-(m-tolyl)formazan ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: m-Tolyltetrazolium Red Formazan

B0283-100MG
TCB0283-100MGEA 32.46 USD
TCB0283-100MG
3,5-Diphenyl-1-(m-tolyl)formazan ≥98.0% (by HPLC)
3,5-Diphenyl-1-(m-tolyl)formazan

CAS Number: 71274-65-2
MDL Number: MFCD00059314
Molecular Formula: C20H18N4
Molecular Weight: 314.39
Purity/Analysis Method: 98.0% (HPLC)
Form: Crystal
Melting point (°C): 156


Other Product Names / Synonyms: m-Tolyltetrazolium Red Formazan

Formula: C₂₀H₁₈N₄
MW: 314.39 g/mol
Melting Pt: 156 °C
MDL Number: MFCD00059314
CAS Number: 71274-65-2
https://us.vwr.com/stibo/web/std.lang.all/23/39/28932339.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR