3,5-Diphenyl-1-(1-naphthyl)formazan ≥97.0% (by HPLC)

Supplier: TCI America

Synonyms: Violet Tetrazolium FormazanTetrazolium Violet Formazan

B0281-100MG B0281-1G
TCB0281-100MGEA 42.23 USD
TCB0281-100MG TCB0281-001G
3,5-Diphenyl-1-(1-naphthyl)formazan ≥97.0% (by HPLC)
3,5-Diphenyl-1-(1-naphthyl)formazan

CAS Number: 1719-72-8
MDL Number: MFCD00059534
Molecular Formula: C23H18N4
Molecular Weight: 350.43
Purity/Analysis Method: 97.0% (HPLC)
Form: Crystal


Other Product Names / Synonyms: Tetrazolium Violet Formazan, Violet Tetrazolium Formazan

Formula: C₂₃H₁₈N₄
MW: 350.43 g/mol
MDL Number: MFCD00059534
CAS Number: 1719-72-8
https://us.vwr.com/stibo/web/std.lang.all/23/38/28932338.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR