Benzylidene-2-naphthylamine ≥98.0% (by total nitrogen basis)
Supplier: TCI America
B0261-25G
TCB0261-025GEA
217.12
USD
TCB0261-025G
Benzylidene-2-naphthylamine ≥98.0% (by total nitrogen basis)
Benzylidene-2-naphthylamine
CAS Number: 891-32-7
MDL Number: MFCD00059551
Molecular Formula: C17H13N
Molecular Weight: 231.30
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 101
Formula:
C₁₇H₁₃N Melting Pt: 101 °C |
MDL Number:
MFCD00059551 CAS Number: 891-32-7 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...