Butyl-4-[(4-Methoxybenzylidene)amino]cinnamate ≥98.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
4-(p-Anisalamino)cinnamic Acid Butyl Ester
CAS Number: 16833-17-3
MDL Number: MFCD00059871
Molecular Formula: C21H23NO3
Molecular Weight: 337.42
Purity/Analysis Method:
Form: Crystal
Product Categories: Materials Chemistry, Liquid Crystal Materials (Optically Functional Materials)
Formula:
C₂₁H₂₃NO₃ MW: 337.42 g/mol |
MDL Number:
MFCD00059871 CAS Number: 16833-17-3 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...