m-Toluidine-4-sulfonic acid ≥95.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
4-Amino-2-methylbenzenesulphonic acid
, 5-Aminotoluene-2-sulphonic acid
, 5-Aminotoluene-2-sulfonic acid
, 3-Methylaniline-4-sulfonic acid
, 4-Amino-2-methylbenzenesulfonic acid
, 3-Methylaniline-4-sulphonic acid
A0329-25G
TCA0329-025GEA
55
USD
TCA0329-025G
m-Toluidine-4-sulfonic acid ≥95.0% (by titrimetric analysis)
m-Toluidine-4-sulfonic acid
CAS Number: 133-78-8
MDL Number: MFCD00035774
Molecular Formula: C7H9NO3S
Molecular Weight: 187.21
Purity/Analysis Method:
Form: Crystal
Other Product Names / Synonyms: 5-Aminotoluene-2-sulfonic Acid, 3-Methylaniline-4-sulfonic Acid, 4-Amino-2-methylbenzenesulfonic Acid
Formula:
C₇H₉NO₃S MW: 187.21 g/mol |
MDL Number:
MFCD00035774 CAS Number: 133-78-8 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...