4'-Butoxybenzylidene-4-cyanoaniline ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 4-(4-Butoxybenzylideneamino)benzonitrile

B0252-1G
TCB0252-001GEA 162 USD
TCB0252-001G
4'-Butoxybenzylidene-4-cyanoaniline ≥98.0% (by titrimetric analysis)
4'-Butoxybenzylidene-4-cyanoaniline

CAS Number: 36405-17-1
MDL Number: MFCD00063742
Molecular Formula: C18H18N2O
Molecular Weight: 278.36
Purity/Analysis Method: 98.0% (T)
Form: Crystal


Product Categories: Materials Chemistry, Liquid Crystal Materials (Optically Functional Materials)
Other Product Names / Synonyms: 4-(4-Butoxybenzylideneamino)benzonitrile

Formula: C₁₈H₁₈N₂O
MW: 278.36 g/mol
MDL Number: MFCD00063742
CAS Number: 36405-17-1
https://us.vwr.com/stibo/web/std.lang.all/23/22/28932322.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR