N-(4-Methoxy-2-hydroxybenzylidene)-4-butylaniline ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
Synonyms:
N-(2-Hydroxy-p-anisal)-4-butylaniline
B0250-1G
TCB0250-001GEA
347
USD
TCB0250-001G
N-(4-Methoxy-2-hydroxybenzylidene)-4-butylaniline ≥98.0% (by HPLC, titration analysis)
N-(4-Methoxy-2-hydroxybenzylidene)-4-butylaniline
CAS Number: 30633-94-4
MDL Number: MFCD00059253
Molecular Formula: C18H21NO2
Molecular Weight: 283.37
Purity/Analysis Method:
Form: Crystal
Product Categories: Materials Chemistry, Liquid Crystal Materials (Optically Functional Materials)
Other Product Names / Synonyms: N-(2-Hydroxy-p-anisal)-4-butylaniline, N-(2-Hydroxy-p-anisylidene)-4-butylaniline
Formula:
C₁₈H₂₁NO₂ MW: 283.37 g/mol |
MDL Number:
MFCD00059253 CAS Number: 30633-94-4 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...