Bis(2,4,5-trichlorophenyl)disulfide ≥95.0%
Supplier: TCI America
Synonyms:
2,4,5-Trichlorophenyl Disulphide
, Di(2,4,5-trichlorophenyl) Disulphide
, 2,4,5-Trichlorophenyl Disulfide
, Di(2,4,5-trichlorophenyl) Disulfide
CAS Number: 3808-87-5
MDL Number: MFCD00000551
Molecular Formula: C12H4Cl6S2
Molecular Weight: 424.98
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 147
Other Product Names / Synonyms: Di(2,4,5-trichlorophenyl) Disulfide, 2,4,5-Trichlorophenyl Disulfide
Formula:
C₁₂H₄Cl₆S₂ MW: 424.98 g/mol Melting Pt: 147 °C |
MDL Number:
MFCD00000551 CAS Number: 3808-87-5 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...