ɑ,ε-Dibenzoyl-DL-lysine ≥99.0% (by titrimetric analysis)

Supplier: TCI America

B0208-1G B0208-5G
TCB0208-001GEA 95.36 USD
TCB0208-001G TCB0208-5G
ɑ,ε-Dibenzoyl-DL-lysine ≥99.0% (by titrimetric analysis)
alpha,epsilon-Dibenzoyl-DL-lysine

CAS Number: 32513-92-1
MDL Number: MFCD00059684
Molecular Formula: C20H22N2O4
Molecular Weight: 354.41
Purity/Analysis Method: 99.0% (T)
Form: Crystal
Melting point (°C): 146

Formula: C₂₀H₂₂N₂O₄
MW: 354.41 g/mol
Melting Pt: 146 °C
MDL Number: MFCD00059684
CAS Number: 32513-92-1
https://us.vwr.com/stibo/web/std.lang.all/22/99/28932299.jpg

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR