N-[Bis(4-methoxyphenyl)methylene]benzylamine ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: Schoenberg's ReagentN-(4,4'-Dimethoxybenzhydrylidene)benzylamine

B0137-1G
TCB0137-001GEA 238 USD
TCB0137-001G
N-[Bis(4-methoxyphenyl)methylene]benzylamine ≥98.0% (by HPLC, titration analysis)
N-[Bis(4-methoxyphenyl)methylene]benzylamine

CAS Number: 524-96-9
MDL Number: MFCD00025824
Molecular Formula: C22H21NO2
Molecular Weight: 331.42
Purity/Analysis Method: 98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 91


Other Product Names / Synonyms: N-(4,4'-Dimethoxybenzhydrylidene)benzylamine, Schoenberg's Reagent

Formula: C₂₂H₂₁NO₂
MW: 331.42 g/mol
Melting Pt: 91 °C
MDL Number: MFCD00025824
CAS Number: 524-96-9
https://us.vwr.com/stibo/web/std.lang.all/22/89/28932289.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR