TCIAB0070

B0070-25ML
TCB0070-025MLEA 279.43 USD
TCB0070-025ML
TCIAB0070
CAS Number: 39252-69-2
MDL Number: MFCD00059491
Molecular Formula: C9H9IO2
Molecular Weight: 276.07
Purity/Analysis Method: 97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 163
Specific Gravity (20/20): 1.67

Other Product Names / Synonyms: Benzoic Acid 2-Iodoethyl Ester
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR