2-Chloroethyl benzoate ≥99.0%
Supplier: TCI America
Synonyms:
Benzoic Acid 2-Chloroethyl Ester
B0068-25ML
TCB0068-025MLEA
137.99
USD
TCB0068-025ML
2-Chloroethyl benzoate ≥99.0%
2-Chloroethyl benzoate
CAS Number: 939-55-9
MDL Number: MFCD00045290
Molecular Formula: C9H9ClO2
Molecular Weight: 184.62
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 257
Specific Gravity (20/20): 1.21
Other Product Names / Synonyms: Benzoic Acid 2-Chloroethyl Ester
Formula:
C₉H₉ClO₂ MW: 184.62 g/mol Boiling Pt: 257 °C Density: 1.21 (20 °C) |
MDL Number:
MFCD00045290 CAS Number: 939-55-9 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...