Nickel(II) benzenesulfonate hexahydrate ≥98.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
Benzenesulfonic Acid Nickel(II) Salt
B0034-25G
TCB0034-025GEA
209
USD
TCB0034-025G
Nickel(II) benzenesulfonate hexahydrate ≥98.0% (by titrimetric analysis)
Nickel(II) benzenesulfonate hexahydrate
CAS Number: 39819-65-3
MDL Number: MFCD00070595
Molecular Formula: C6H6O3S
Molecular Weight: 373.02
Purity/Analysis Method:
Form: Crystal
Product Categories: Synthetic Organic Chemistry, Activation Reaction of Unactivated Bonds
Other Product Names / Synonyms: Benzenesulfonic Acid Nickel(II) Salt
Formula:
C₆H₆O₃S MW: 373.02 g/mol |
MDL Number:
MFCD00070595 CAS Number: 39819-65-3 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...