DBD-F [=4-(N,N-dimethylaminosulfonyl)-7-fluoro-2,1,3-benzoxadiazole] ≥98.0% (by HPLC)
Supplier: TCI America
Synonyms:
4-(N,N-Dimethylaminosulfonyl)-7-fluoro-2,1,3-benzoxadiazole
[for HPLC Labeling]
CAS Number: 98358-90-8
MDL Number: MFCD00143356
Molecular Formula: C8H8FN3O3S
Molecular Weight: 245.23
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 128
Storage Temperature: 0-10°C
Product Categories: Bioscience, Fluorescent Labeling Dyes, Analytical Science, Fluorescent Dyes, Fluorescence Detection (HPLC Labeling Reagents), Fluorescence Detection (HPLC Labeling Reagents)
Other Product Names / Synonyms: 4-(N,N-Dimethylaminosulfonyl)-7-fluoro-2,1,3-benzoxadiazole, 7-Fluoro-4-(N,N-dimethylaminosulfonyl)benzofurazan, 7-Fluoro-4-(N,N-dimethylsulfamoyl)-2,1,3-benzoxadiazole
Formula:
C₈H₈FN₃O₃S MW: 245.23 g/mol Melting Pt: 128 °C |
MDL Number:
MFCD00143356 CAS Number: 98358-90-8 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...