4-(4,5-Diphenyl-1H-imidazol-2-yl)benzoyl chloride hydrochloride ≥97.0% (by titrimetric analysis)
Supplier: TCI America
A5579-100MG
TCA5579-100MGEA
123
USD
TCA5579-100MG
4-(4,5-Diphenyl-1H-imidazol-2-yl)benzoyl chloride hydrochloride ≥97.0% (by titrimetric analysis)
4-(4,5-Diphenyl-1H-imidazol-2-yl)benzoyl chloride hydrochloride
[for HPLC Labeling]
MDL Number: MFCD03844764
Molecular Formula: C22H15ClN2O
Molecular Weight: 395.28
Purity/Analysis Method:
Form: Crystal
Storage Temperature: 0-10°C
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...