AABD-SH (=4-Acetamido-7-mercapto-2,1,3-benzoxadiazole) ≥95.0% (by HPLC)
Supplier: TCI America
Synonyms:
4-Acetamido-7-mercaptobenzofurazan
, 4-Acetamido-7-mercapto-2,1,3-benzoxadiazole
A5576-100MG
TCA5576-100MGEA
321.63
USD
TCA5576-100MG
AABD-SH (=4-Acetamido-7-mercapto-2,1,3-benzoxadiazole) ≥95.0% (by HPLC)
AABD-SH (=4-Acetamido-7-mercapto-2,1,3-benzoxadiazole)
[for HPLC Labeling]
CAS Number: 254973-02-9
MDL Number: MFCD04038408
Molecular Formula: C8H7N3O2S
Molecular Weight: 209.22
Purity/Analysis Method:
Form: Crystal
Storage Temperature: 0-10°C
Product Categories: Analytical Science, Fluorescence Detection (HPLC Labeling Reagents)
Other Product Names / Synonyms: 4-Acetamido-7-mercaptobenzofurazan, 4-Acetamido-7-mercapto-2,1,3-benzoxadiazole
Formula:
C₈H₇N₃O₂S MW: 209.22 g/mol |
MDL Number:
MFCD04038408 CAS Number: 254973-02-9 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...