DBD-NCS [=4-(N,N-dimethylaminosulfonyl)-7-isothiocyanato-2,1,3-benzoxadiazole]
Supplier: TCI America
Synonyms:
4-(N,N-Dimethylaminosulfonyl)-7-isothiocyanatobenzofurazan
[for HPLC Labeling and Edman Degradation]
CAS Number: 147611-81-2
MDL Number: MFCD03844763
Molecular Formula: C9H8N4O3S2
Molecular Weight: 284.31
Form: Crystal
Color: Slightly Pale Yellow
Melting point (°C): 126
Storage Temperature: 0-10°C
Other Product Names / Synonyms: 4-(N,N-Dimethylaminosulfonyl)-7-isothiocyanatobenzofurazan, 4-(N,N-Dimethylaminosulfonyl)-7-isothiocyanato-2,1,3-benzoxadiazole
Formula:
C₉H₈N₄O₃S₂ MW: 284.31 g/mol Melting Pt: 126 °C |
MDL Number:
MFCD03844763 CAS Number: 147611-81-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...