(S)-(+)-DBD-Py-NCS [=(S)-(+)-4-(N,N-Dimethylaminosulfonyl)-7-(3-isothiocyanatopyrrolidin-1-yl)-2,1,3-benzoxadiazole] ≥98.0% (by HPLC)
Supplier: TCI America
Synonyms:
(S)-(+)-4-(N,N-Dimethylaminosulfonyl)-7-(3-isothiocyanatopyrrolidin-1-yl)benzofurazan
[for HPLC Labeling]
CAS Number: 163927-32-0
MDL Number: MFCD00671494
Molecular Formula: C13H15N5O3S2
Molecular Weight: 353.42
Purity/Analysis Method:
Form: Crystal
Specific rotation [a]20/D: 285 deg (C=0.4, CH3CN)
Storage Temperature: 0-10°C
Product Categories: Analytical Science, e.e. Determination (HPLC Labeling Reagents), Fluorescence Detection (HPLC Labeling Reagents), Fluorescence Detection (HPLC Labeling Reagents)
Other Product Names / Synonyms: (S)-(+)-4-(N,N-Dimethylaminosulfonyl)-7-(3-isothiocyanatopyrrolidin-1-yl)benzofurazan, (S)-(+)-4-(N,N-Dimethylaminosulfonyl)-7-(3-isothiocyanatopyrrolidin-1-yl)-2,1,3-benzoxadiazole
Formula:
C₁₃H₁₅N₅O₃S₂ MW: 353.42 g/mol |
MDL Number:
MFCD00671494 CAS Number: 163927-32-0 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...