p-Xylylenebis(N,N-diethyldithiocarbamate) ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

Synonyms: 1,4-Bis(N,N-diethyldithiocarbamylmethyl)benzenep-Xylenebis(N,N-diethyldithiocarbamate)

X0056-5G
TCX0056-005GEA 399.74 USD
TCX0056-005G
p-Xylylenebis(N,N-diethyldithiocarbamate) ≥98.0% (by HPLC, total nitrogen)
p-Xylylenebis(N,N-diethyldithiocarbamate)

CAS Number: 89964-93-2
MDL Number: MFCD00191696
Molecular Formula: C18H28N2S4
Molecular Weight: 400.68
Purity/Analysis Method: 98.0% (HPLC,N)
Form: Crystal
Color: White
Melting point (°C): 79


Other Product Names / Synonyms: 1,4-Bis(N,N-diethyldithiocarbamylmethyl)benzene, p-Xylenebis(N,N-diethyldithiocarbamate)

Formula: C₁₈H₂₈N₂S₄
MW: 400.68 g/mol
Melting Pt: 79 °C
MDL Number: MFCD00191696
CAS Number: 89964-93-2
https://us.vwr.com/stibo/web/std.lang.all/74/04/28997404.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR