2,4-Dimethylaniline-5-sulfonic acid sodium salt hydrate
Supplier: TCI America
Synonyms:
m-Xylidine-5-sulfonic Acid Sodium Salt
, m-Xylidine-5-sulphonic Acid Sodium Salt
X0030-25G
TCX0030-025GEA
449.58
USD
TCX0030-025G
2,4-Dimethylaniline-5-sulfonic acid sodium salt hydrate
2,4-Dimethylaniline-5-sulfonic acid sodium salt hydrate
CAS Number: 64501-84-4
MDL Number: MFCD00070550
Molecular Formula: C8H11NO3S
Molecular Weight: 223.22
Form: Crystal
Color: White
Other Product Names / Synonyms: m-Xylidine-5-sulfonic Acid Sodium Salt
Formula:
C₈H₁₁NO₃S MW: 223.22 g/mol |
MDL Number:
MFCD00070550 CAS Number: 64501-84-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...