2,6-Xylidine hydrochloride ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

Synonyms: 2,6-Dimethylaniline hydrochloride

X0029-25G
TCX0029-025GEA 169 USD
TCX0029-025G
2,6-Xylidine hydrochloride ≥98.0% (by HPLC, total nitrogen)
2,6-Xylidine hydrochloride

CAS Number: 21436-98-6
MDL Number: MFCD00060213
Molecular Formula: C8H11N
Molecular Weight: 157.64
Purity/Analysis Method: 98.0% (HPLC,N)
Form: Crystal
Color: White


Other Product Names / Synonyms: 2,6-Xylidine Hydrochloride

Formula: C₈H₁₂ClN
MW: 157.6406 g/mol
MDL Number: MFCD00060213
CAS Number: 21436-98-6
https://us.vwr.com/stibo/web/std.lang.all/73/87/28997387.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR