2,4-Xylidine hydrochloride ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
Synonyms:
2,4-Dimethylaniline hydrochloride
X0028-25G
TCX0028-025GEA
376
USD
TCX0028-025G
2,4-Xylidine hydrochloride ≥98.0% (by HPLC, titration analysis)
2,4-Xylidine hydrochloride
CAS Number: 21436-96-4
MDL Number: MFCD00060209
Molecular Formula: C8H11N
Molecular Weight: 157.64
Purity/Analysis Method:
Form: Crystal
Color: White
Melting point (°C): 238
Other Product Names / Synonyms: m-Xylidine Hydrochloride
Formula:
C₈H₁₂ClN MW: 157.6406 g/mol Melting Pt: 238 °C |
MDL Number:
MFCD00060209 CAS Number: 21436-96-4 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...