Vitamin K4 ≥98.0%

Supplier: TCI America

Synonyms: Menadiol Diacetate2-Methylnaphthalene-1,4-diyl diacetate1,4-Diacetoxy-2-methylnaphthalene

V0103-25G
TCV0103-25GEA 68 USD
TCV0103-25G
Vitamin K4 ≥98.0%
Vitamin K4

CAS Number: 573-20-6
MDL Number: MFCD00021469
Molecular Formula: C15H14O4
Molecular Weight: 258.27
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Color: White
Melting point (°C): 113


Product Categories: Bioscience, Vitamins
Other Product Names / Synonyms: 1,4-Diacetoxy-2-methylnaphthalene, Menadiol Diacetate

Formula: C₁₅H₁₄O₄
MW: 258.27 g/mol
Melting Pt: 113 °C
MDL Number: MFCD00021469
CAS Number: 573-20-6
https://us.vwr.com/stibo/web/std.lang.all/73/20/28997320.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR