(S)-(-)-NBD-Pro-COCl [=(S)-(-)-4-Nitro-7-(2-chloroformylpyrrolidin-1-yl)-2,1,3-benzoxadiazole]

Supplier: TCI America

Synonyms: (S)-(-)-4-Nitro-7-(2-chloroformylpyrrolidin-1-yl)benzofurazan(S)-(-)-4-Nitro-7-(2-chloroformylpyrrolidin-1-yl)-2,1,3-benzoxadiazole

A5567-100MG
TCA5567-100MGEA 139 USD
TCA5567-100MG
(S)-(-)-NBD-Pro-COCl [=(S)-(-)-4-Nitro-7-(2-chloroformylpyrrolidin-1-yl)-2,1,3-benzoxadiazole]
(S)-(-)-NBD-Pro-COCl [=(S)-(-)-4-Nitro-7-(2-chloroformylpyrrolidin-1-yl)-2,1,3-benzoxadiazole]

[HPLC Labeling Reagent for e.e. Determination]
CAS Number: 159717-68-7
MDL Number: MFCD00191512
Molecular Formula: C11H9ClN4O4
Molecular Weight: 296.67
Form: Crystal
Color: Yellow
Storage Temperature: 0-10°C


Other Product Names / Synonyms: (S)-(-)-4-Nitro-7-(2-chloroformylpyrrolidin-1-yl)benzofurazan, (S)-(-)-4-Nitro-7-(2-chloroformylpyrrolidin-1-yl)-2,1,3-benzoxadiazole

Formula: C₁₁H₉ClN₄O₄
MW: 296.67 g/mol
MDL Number: MFCD00191512
CAS Number: 159717-68-7
https://us.vwr.com/stibo/web/std.lang.all/21/38/28932138.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR