TCIAV0074

V0074-1G
TCV0074-001GEA 391.73 USD
TCV0074-001G
TCIAV0074
CAS Number: 60081-73-4
MDL Number: MFCD01861277
Molecular Formula: C8H8O2S
Molecular Weight: 190.19
Form: Crystal
Color: Pale Yellow
Storage Temperature: 0-10°C

Other Product Names / Synonyms: 4-Vinylbenzenesulfinic Acid Sodium Salt
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR