4-Hydroxy-5-decanone ≥97.0%

Synonyms: Valeroin6-Hydroxy-5-decanone

V0055-5ML V0055-25ML
TCV0055-025MLEA 510.88 USD
TCV0055-025ML TCV0055-5ML
4-Hydroxy-5-decanone ≥97.0%
4-Hydroxy-5-decanone

CAS Number: 6540-98-3
MDL Number: MFCD00059663
Molecular Formula: C10H20O2
Molecular Weight: 172.27
Purity/Analysis Method: 97.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 92
Specific Gravity (20/20): 0.91


Other Product Names / Synonyms: 6-Hydroxy-5-decanone

Formula: C₁₀H₂₀O₂
MW: 172.27 g/mol
Boiling Pt: 92 °C
Density: 0.91 (20 °C)
MDL Number: MFCD00059663
CAS Number: 6540-98-3
EINECS: 229-455-5
https://us.vwr.com/stibo/web/std.lang.all/72/93/28997293.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR