Valeryl bromide ≥98.0%

Supplier: TCI America

Synonyms: Valeroyl Bromide

V0038-10G
TCV0038-010GEA 224.25 USD
TCV0038-010G
Valeryl bromide ≥98.0%
Valeryl bromide

CAS Number: 1889-26-5
MDL Number: MFCD00059467
Molecular Formula: C5H9BrO
Molecular Weight: 165.03
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 64
Specific Gravity (20/20): 1.34


Other Product Names / Synonyms: Valeroyl Bromide

Formula: C₅H₉BrO
MW: 165.03 g/mol
Boiling Pt: 64 °C
Density: 1.34 (20 °C)
MDL Number: MFCD00059467
CAS Number: 1889-26-5

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR