(±)-4-Vinylcyclohexene ≥95.0% stabilized
Supplier: TCI America
Synonyms:
(±)-4-Ethenyl-1-cyclohexene
, 4-Vinyl-1-cyclohexene
V0023-25ML
TCV0023-025MLEA
54.1
USD
TCV0023-025ML
(±)-4-Vinylcyclohexene ≥95.0% stabilized
(±)-4-Vinylcyclohexene
(stabilized with BHT)
CAS Number: 100-40-3
MDL Number: MFCD00001576
Molecular Formula: C8H12
Molecular Weight: 108.18
Purity/Analysis Method:
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 128
Flash Point (°C): 16
Specific Gravity (20/20): 0.83
Formula:
C₈H₁₂ MW: 108.18 g/mol Boiling Pt: 128 °C Melting Pt: –108.7 °C Density: 0.83 (20 °C) Flash Pt: 16 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00001576 CAS Number: 100-40-3 EINECS: 202-848-9 UN: 3295 ADR: 3,II |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...