9-Vinylcarbazole ≥98.0%

Supplier: TCI America

Synonyms: N-Vinylcarbazole

V0021-25G
TCV0021-025GEA 61.33 USD
TCV0021-025G
9-Vinylcarbazole ≥98.0%
9-Vinylcarbazole

CAS Number: 1484-13-5
MDL Number: MFCD00004966
Molecular Formula: C14H11N
Molecular Weight: 193.25
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Color: White
Boiling point (°C): 180
Melting point (°C): 65
Flash Point (°C): 182

Formula: C₁₄H₁₁N
MW: 193.25 g/mol
Boiling Pt: 180 °C
Melting Pt: 65 °C
Flash Pt: 182 °C
MDL Number: MFCD00004966
CAS Number: 1484-13-5
https://us.vwr.com/stibo/web/std.lang.all/72/72/28997272.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR