(S)-(-)-DBD-Pro-COCl [=(S)-(-)-4-(N,N-Dimethylaminosulfonyl)-7-(2-chloroformylpyrrolidin-1-yl)-2,1,3-benzoxadiazole] ≥90.0% (by HPLC)
Supplier: TCI America
Synonyms:
(S)-(-)-4-(N,N-Dimethylaminosulfonyl)-7-(2-chloroformylpyrrolidin-1-yl)benzofurazan
[HPLC Labeling Reagent for e.e. Determination]
CAS Number: 150993-63-8
MDL Number: MFCD00191380
Molecular Formula: C13H15ClN4O4S
Molecular Weight: 358.80
Purity/Analysis Method:
Form: Crystal
Storage Temperature: 0-10°C
Product Categories: Analytical Science, e.e. Determination (HPLC Labeling Reagents), Fluorescence Detection (HPLC Labeling Reagents), Fluorescence Detection (HPLC Labeling Reagents)
Other Product Names / Synonyms: (S)-(-)-4-(N,N-Dimethylaminosulfonyl)-7-(2-chloroformylpyrrolidin-1-yl)benzofurazan, (S)-(-)-4-(N,N-Dimethylaminosulfonyl)-7-(2-chloroformylpyrrolidin-1-yl)-2,1,3-benzoxadiazole
Formula:
C₁₃H₁₅ClN₄O₄S MW: 358.80 g/mol |
MDL Number:
MFCD00191380 CAS Number: 150993-63-8 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...