3-Undecylthiophene ≥97.0%

Supplier: TCI America

U0056-5G U0056-1G
TCU0056-001GEA 179 USD
TCU0056-001G TCU0056-005G
3-Undecylthiophene ≥97.0%
3-Undecylthiophene

CAS Number: 129607-86-9
MDL Number: MFCD00130144
Molecular Formula: C15H26S
Molecular Weight: 238.43
Purity/Analysis Method: 97.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 166
Specific Gravity (20/20): 0.91

Formula: C₁₅H₂₆S
MW: 238.4300 g/mol
Boiling Pt: 166 °C
Density: 0.91 (20 °C)
MDL Number: MFCD00130144
CAS Number: 129607-86-9
https://us.vwr.com/stibo/web/std.lang.all/72/17/28997217.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR