4-Undecanol ≥98.0%
Supplier: TCI America
U0038-5ML
TCU0038-005MLEA
227.25
USD
TCU0038-005ML
4-Undecanol ≥98.0%
4-Undecanol
CAS Number: 4272-06-4
MDL Number: MFCD00046724
Molecular Formula: C11H24O
Molecular Weight: 172.31
Purity/Analysis Method:
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 80
Flash Point (°C): 66
Specific Gravity (20/20): 0.83
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...