1,10-Undecadiene ≥90.0%

Supplier: TCI America

U0024-5ML U0024-1ML
TCU0024-005MLEA 544.89 USD
TCU0024-005ML TCU0024-1ML
1,10-Undecadiene ≥90.0%
1,10-Undecadiene

CAS Number: 13688-67-0
MDL Number: MFCD00059203
Molecular Formula: C11H20
Molecular Weight: 152.28
Purity/Analysis Method: 90.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 187
Flash Point (°C): 38
Specific Gravity (20/20): 0.77

Formula: C₁₁H₂₀
Boiling Pt: 187 °C
Density: 0.77 (20 °C)
Flash Pt: 38 °C
MDL Number: MFCD00059203
CAS Number: 13688-67-0
https://us.vwr.com/stibo/web/std.lang.all/71/86/28997186.jpg

Order Now

Specification Test Results

Purity/Analysis Method 90.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR