TCIAU0022

U0022-100MG
TCU0022-100MGEA 47.88 USD
TCU0022-100MG
TCIAU0022
CAS Number: 19817-92-6
MDL Number: MFCD00044310
Molecular Formula: C9H15N2O15P3
Molecular Weight: 550.08
Purity/Analysis Method: 95.0% (HPLC)
Form: Crystal
Color: White
Flash Point (°C): 113
Storage Temperature: 0°C

Product Categories: Bioscience, Nucleotides and Their Analogs
Other Product Names / Synonyms: 5'-UTP 3Na
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR