Tetrakis(acetonitrile)copper(I) hexafluorophosphate ≥97.0% (by titrimetric analysis)
Supplier: TCI America
T2665-5G
TCT2665-5GEA
64.2
USD
TCT2665-5G
Tetrakis(acetonitrile)copper(I) hexafluorophosphate ≥97.0% (by titrimetric analysis)
Tetrakis(acetonitrile)copper(I) hexafluorophosphate
CAS Number: 64443-05-6
MDL Number: MFCD00064810
Molecular Formula: C8H12CuF6N4P
Molecular Weight: 372.72
Purity/Analysis Method:
Form: Crystal
Color: White
Melting point (°C): 160
Storage Temperature:
Formula:
C₈H₁₂CuF₆N₄P Melting Pt: 160 °C |
MDL Number:
MFCD00064810 CAS Number: 64443-05-6 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...