DBD-COCl [=4-(N,N-dimethylaminosulfonyl)-7-(N-chloroformylmethyl-N-methylamino)-2,1,3-benzoxadiazole] ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 4-(N,N-Dimethylaminosulfonyl)-7-(N-chloroformylmethyl-N-methylamino)benzofurazan4-(N,N-Dimethylaminosulfonyl)-7-(N-chloroformylmethyl-N-methylamino)-2,1,3-benzoxadiazole

A5558-100MG
TCA5558-100MGEA 295 USD
TCA5558-100MG
DBD-COCl [=4-(N,N-dimethylaminosulfonyl)-7-(N-chloroformylmethyl-N-methylamino)-2,1,3-benzoxadiazole] ≥98.0% (by titrimetric analysis)
DBD-COCl [=4-(N,N-dimethylaminosulfonyl)-7-(N-chloroformylmethyl-N-methylamino)-2,1,3-benzoxadiazole]

[for HPLC Labeling]
CAS Number: 156153-43-4
MDL Number: MFCD00191377
Molecular Formula: C11H13ClN4O4S
Molecular Weight: 332.76
Purity/Analysis Method: 98.0% (T)
Form: Crystal
Storage Temperature: 0°C


Product Categories: Analytical Science, Fluorescence Detection (HPLC Labeling Reagents), Fluorescence Detection (HPLC Labeling Reagents), Fluorescence Detection (HPLC Labeling Reagents)
Other Product Names / Synonyms: 4-(N,N-Dimethylaminosulfonyl)-7-(N-chloroformylmethyl-N-methylamino)benzofurazan, 4-(N,N-Dimethylaminosulfonyl)-7-(N-chloroformylmethyl-N-methylamino)-2,1,3-benzoxadiazole

Formula: C₁₁H₁₃ClN₄O₄S
MW: 332.76 g/mol
MDL Number: MFCD00191377
CAS Number: 156153-43-4
https://us.vwr.com/stibo/web/std.lang.all/21/30/28932130.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR