(R)-(-)-2,3,7,7a-Tetrahydro-7a-methyl-1H-indene-1,5(6H)-dione ≥98.0%

Synonyms: (-)-Hajos-Parrish Ketone

T2636-1G
TCT2636-1GEA 266.77 USD
TCT2636-1G
(R)-(-)-2,3,7,7a-Tetrahydro-7a-methyl-1H-indene-1,5(6H)-dione ≥98.0%
(R)-(-)-2,3,7,7a-Tetrahydro-7a-methyl-1H-indene-1,5(6H)-dione

CAS Number: 17553-89-8
MDL Number: MFCD08235115
Molecular Formula: C10H12O2
Molecular Weight: 164.20
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Color: Very Pale Yellow
Melting point (°C): 59
Specific rotation [a]20/D: -299 deg (C=1, Toluene)


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: (-)-Hajos-Parrish Ketone

Formula: C₁₀H₁₂O₂
MW: 164.20 g/mol
Melting Pt: 59 °C
MDL Number: MFCD08235115
CAS Number: 17553-89-8
https://us.vwr.com/stibo/web/std.lang.all/63/45/28996345.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR