(Tetrahydrofuran-2-yl)acetonitrile ≥98.0%
Supplier: TCI America
Synonyms:
2-(Cyanomethyl)tetrahydrofuran
, Tetrahydrofurfurylcarbonitrile
, 2-(Tetrahydrofuran-2-yl)acetonitrile
CAS Number: 33414-62-9
MDL Number: MFCD03206782
Molecular Formula: C6H9NO
Molecular Weight: 111.14
Purity/Analysis Method:
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 92
Specific Gravity (20/20): 1.02
Other Product Names / Synonyms: Tetrahydrofurfurylcarbonitrile, 2-(Cyanomethyl)tetrahydrofuran
Formula:
C₆H₉NO MW: 111.14 g/mol Boiling Pt: 92 °C Density: 1.02 (20 °C) |
MDL Number:
MFCD03206782 CAS Number: 33414-62-9 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...