DBD-H [=4-(N,N-dimethylaminosulfonyl)-7-hydrazino-2,1,3-benzoxadiazole] ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: 4-(N,N-Dimethylaminosulfonyl)-7-hydrazinobenzofurazan4-(N,N-Dimethylaminosulfonyl)-7-hydrazino-2,1,3-benzoxadiazole

A5556-100MG
TCA5556-100MGEA 299.34 USD
TCA5556-100MG
DBD-H [=4-(N,N-dimethylaminosulfonyl)-7-hydrazino-2,1,3-benzoxadiazole] ≥98.0% (by HPLC)
DBD-H [=4-(N,N-dimethylaminosulfonyl)-7-hydrazino-2,1,3-benzoxadiazole]

[for HPLC Labeling]
CAS Number: 131467-86-2
MDL Number: MFCD00191376
Molecular Formula: C8H11N5O3S
Molecular Weight: 257.27
Purity/Analysis Method: 98.0% (HPLC)
Form: Crystal
Storage Temperature: 0°C


Product Categories: Analytical Science, Fluorescence Detection (HPLC Labeling Reagents)
Other Product Names / Synonyms: 4-(N,N-Dimethylaminosulfonyl)-7-hydrazinobenzofurazan, 4-(N,N-Dimethylaminosulfonyl)-7-hydrazino-2,1,3-benzoxadiazole

Formula: C₈H₁₁N₅O₃S
MW: 257.27 g/mol
MDL Number: MFCD00191376
CAS Number: 131467-86-2
https://us.vwr.com/stibo/web/std.lang.all/21/28/28932128.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR