5,10,15,20-Tetrakis[3,5-bis(per-O-methyl-α-cyclodextrin-6-yloxy)phenyl]-21H,23H-porphine

Supplier: TCI America

Synonyms: 5,10,15,20-Tetrakis[3,5-bis(per-O-methyl-α-cyclodextrin-6-yloxy)phenyl]porphyrin

T2452-10MG
TCT2452-10MGEA 399 USD
TCT2452-10MG
5,10,15,20-Tetrakis[3,5-bis(per-O-methyl-α-cyclodextrin-6-yloxy)phenyl]-21H,23H-porphine
5,10,15,20-Tetrakis[3,5-bis(per-O-methyl-alpha-cyclodextrin-6-yloxy)phenyl]-21H,23H-porphine

Molecular Weight: 10289.06
Form: Crystal
Color: Slightly Pale Purple
Storage Temperature: 0-10°C
Molecular Formula: C468H766N4O240


Other Product Names / Synonyms: 5,10,15,20-Tetrakis[3,5-bis(per-O-methyl-alpha-cyclodextrin-6-yloxy)phenyl]porphyrin

Formula: C₄₆₈H₇₆₆N₄O₂₄₀
MW: 10288.97 g/mol
CAS Number: 117194-77-1
https://us.vwr.com/stibo/web/std.lang.all/61/92/28996192.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR