5,10,15,20-Tetrakis[4-(per-O-methyl-α-cyclodextrin-6-yloxy)phenyl]porphyrin

Supplier: TCI America

Synonyms: 5,10,15,20-Tetrakis[4-(per-O-methyl-α-cyclodextrin-6-yloxy)phenyl]-21H,23H-porphine

T2451-10MG
TCT2451-10MGEA 399 USD
TCT2451-10MG
5,10,15,20-Tetrakis[4-(per-O-methyl-α-cyclodextrin-6-yloxy)phenyl]porphyrin
5,10,15,20-Tetrakis[4-(per-O-methyl-alpha-cyclodextrin-6-yloxy)phenyl]porphyrin

Molecular Weight: 5451.91
Form: Crystal
Color: Deep Red
Storage Temperature: 0-10°C
Molecular Formula: C256H398N4O120


Other Product Names / Synonyms: 5,10,15,20-Tetrakis[4-(per-O-methyl-alpha-cyclodextrin-6-yloxy)phenyl]-21H,23H-porphine

https://us.vwr.com/stibo/web/std.lang.all/61/91/28996191.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR