2-p-Terphenylboronic Acid (contains varying amounts of Anhydride)
Supplier: TCI America
T2412-1G
TCT2412-1GEA
145
USD
TCT2412-1G
2-p-Terphenylboronic Acid (contains varying amounts of Anhydride)
2-p-Terphenylboronic acid
CAS Number: 663954-31-2
Molecular Formula: C18H15BO2
Molecular Weight: 274.13
Form: Crystal
Color: White
Formula:
C₁₈H₁₅BO₂ |
CAS Number:
663954-31-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...