2,4-Dinitrophenylhydrazine hydrochloride ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

A5531-5G
TCA5531-005GEA 79.76 USD
TCA5531-005G
2,4-Dinitrophenylhydrazine hydrochloride ≥98.0% (by HPLC, titration analysis)
2,4-Dinitrophenylhydrazine hydrochloride

[for HPLC Labeling]
CAS Number: 55907-61-4
MDL Number: MFCD00050688
Molecular Formula: C6H6N4O4
Molecular Weight: 234.60
Purity/Analysis Method: 98.0% (HPLC,T)
Form: Crystal

Formula: C₆H₆N₄O₄
MW: 234.60 g/mol
MDL Number: MFCD00050688
CAS Number: 55907-61-4
https://us.vwr.com/stibo/web/std.lang.all/21/21/28932121.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR