D-ɑ-Tocopherol acetate ≥96.0%

Supplier: TCI America

Synonyms: Vitamin E Acetate (D-form)(+)-ɑ-Tocopherol Acetate

T2322-25G
TCT2322-25GEA 212.09 USD
TCT2322-25G
D-ɑ-Tocopherol acetate ≥96.0%
D-alpha-Tocopherol acetate

CAS Number: 58-95-7
Molecular Formula: C31H52O3
Molecular Weight: 472.75
Purity/Analysis Method: 96.0% (GC)
Form: Clear Liquid
Color: Colorless
Flash Point (°C): 245
Specific rotation [a]20/D: 24 deg


Product Categories: Bioscience, Antioxidants, Vitamins, Terpenes (Others)
Other Product Names / Synonyms: Vitamin E Acetate (D-form)

Formula: C₃₁H₅₂O₃
MW: 472.75 g/mol
Flash Pt: 245 °C
MDL Number: MFCD00072052
CAS Number: 58-95-7
https://us.vwr.com/stibo/web/std.lang.all/61/02/28996102.jpg

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR