D-ɑ-Tocopherylquinone ≥97.0% (by HPLC)

Supplier: TCI America

Synonyms: Vitamin E Quinone (D-form)

T2283-500MG
TCT2283-500MGEA 332 USD
TCT2283-500MG
D-ɑ-Tocopherylquinone ≥97.0% (by HPLC)
D-alpha-Tocopherylquinone

CAS Number: 7559-04-8
Molecular Formula: C29H50O3
Molecular Weight: 446.72
Purity/Analysis Method: 97.0% (HPLC)
Form: Clear Liquid
Color: Colorless
Storage Temperature: 0-10°C


Product Categories: Bioscience, Vitamins, Terpenes (Others)
Other Product Names / Synonyms: Vitamin E Quinone (D-form)

Formula: C₂₉H₅₀O₃
MW: 446.72 g/mol
MDL Number: MFCD00135657
CAS Number: 7559-04-8
https://us.vwr.com/stibo/web/std.lang.all/60/70/28996070.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR