Tetrabutylphosphonium tetraphenylborate ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
T2008-25G
TCT2008-25GEA
135.95
USD
TCT2008-25G
Tetrabutylphosphonium tetraphenylborate ≥98.0% (by HPLC, titration analysis)
Tetrabutylphosphonium tetraphenylborate
CAS Number: 29089-62-1
MDL Number: MFCD04038112
Molecular Formula: C40H56BP
Molecular Weight: 578.67
Purity/Analysis Method:
Form: Crystal
Color: White
Melting point (°C): 230
Formula:
C₄₀H₅₆BP MW: 578.6600 g/mol Melting Pt: 230 °C |
MDL Number:
MFCD04038112 CAS Number: 29089-62-1 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...