D(+)-α-Acetofluoroglucose ≥98.0%
Supplier: TCI America
Synonyms:
2,3,4,6-Tetra-O-acetyl-alpha-D-glucopyranosyl Fluoride
T1995-1G
TCT1995-1GEA
297.26
USD
TCT1995-1G
D(+)-α-Acetofluoroglucose ≥98.0%
D(+)-α-Acetofluoroglucose
CAS Number: 3934-29-0
MDL Number: MFCD00792705
Molecular Formula: C14H19FO9
Molecular Weight: 350.30
Purity/Analysis Method:
Form: Crystal
Color: White
Storage Temperature:
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...