N-(2,4,6-Trichlorophenyl)maleimide ≥98.0% (by HPLC)
Supplier: TCI America
T1987-25G
TCT1987-25GEA
148.13
USD
TCT1987-25G
N-(2,4,6-Trichlorophenyl)maleimide ≥98.0% (by HPLC)
N-(2,4,6-Trichlorophenyl)maleimide
CAS Number: 13167-25-4
MDL Number: MFCD06797173
Molecular Formula: C10H4Cl3NO2
Molecular Weight: 276.50
Purity/Analysis Method:
Form: Crystal
Color: White
Melting point (°C): 134
Formula:
C₁₀H₄Cl₃NO₂ Melting Pt: 134 °C |
MDL Number:
MFCD06797173 CAS Number: 13167-25-4 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...