2,3,5,6-Tetrafluoro-4-(trifluoromethyl)phenol ≥97.0%

Supplier: TCI America

Synonyms: 4-Hydroxy-2,3,5,6-tetrafluorobenzotrifluoride

T1983-5G
TCT1983-5GEA 190.3 USD
TCT1983-5G
2,3,5,6-Tetrafluoro-4-(trifluoromethyl)phenol ≥97.0%
2,3,5,6-Tetrafluoro-4-(trifluoromethyl)phenol

CAS Number: 2787-79-3
MDL Number: MFCD00155984
Molecular Formula: C7HF7O
Molecular Weight: 234.07
Purity/Analysis Method: 97.0% (GC)
Form: Crystal
Color: White
Flash Point (°C): 82
Freezing point (°C): 27


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks, Fluorinated Building Blocks
Other Product Names / Synonyms: alpha,alpha,alpha,2,3,5,6-Heptafluoro-p-cresol, Perfluoro-p-cresol, 2,3,5,6-Tetrafluoro-4-hydroxybenzotrifluoride

Formula: C₇HF₇O
Flash Pt: 82 °C
MDL Number: MFCD00155984
CAS Number: 2787-79-3
https://us.vwr.com/stibo/web/std.lang.all/59/28/28995928.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR